Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethylquinolinium iodide, 98%

$272.73 - $272.73

Chemical Identifiers

Chemical Name or Material 1-Ethylquinolinium iodide
CAS 634-35-5
Molecular Formula C11H12IN
Formula Weight 285.13
MDL Number MFCD00041996
Synonym Quinoline ethiodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1785422

  SDS
alfa aesar™
A1785422
100g Each for $272.73
Description & Specifications

Specifications

Formula Weight 285.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Ethylquinolinium iodide
Molecular Formula C11H12IN
RTECS Number VC3677000
MDL Number MFCD00041996
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Melting Point 156° to 159°C
Quantity 100g
TSCA TSCA
Synonym Quinoline ethiodide
Assay 98%
CAS 634-35-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.