missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylpyrrolidine, TCI America™

Manufacturer:  TCI America E051025G

 View more versions of this product

Catalog No. E051025G



Specifications

Specifications

1-Ethylpyrrolidine
Yellow
7335-06-0
MFCD01632186
ONQBOTKLCMXPOF-UHFFFAOYSA-N
1-ethylpyrrolidine
81782
Clear Liquid at 20°C
105°C
25g
C6H13N
2733
CCN1CCCC1
99.177
99.18
≥98.0% (GC)

Chemical Identifiers

7335-06-0
99.177
ONQBOTKLCMXPOF-UHFFFAOYSA-N
1-ethylpyrrolidine
C6H13N
MFCD01632186
81782
CCN1CCCC1
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6747

TSCA : No

Documents