missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylpyrrole, TCI America™

Manufacturer:  TCI America E04441G

 View more versions of this product

Catalog No. E04441G



Specifications

Specifications

1-Ethylpyrrole
Undesignated
617-92-5
MFCD00060633
VPUAYOJTHRDUTK-UHFFFAOYSA-N
1-ethylpyrrole
185226
Clear Liquid at 20°C
131°C
1g
C6H9N
1993
CCN1C=CC=C1
95.145
95.15
≥98.0% (GC)

Chemical Identifiers

617-92-5
95.145
VPUAYOJTHRDUTK-UHFFFAOYSA-N
1-ethylpyrrole
C6H9N
MFCD00060633
185226
CCN1C=CC=C1
Safety and Handling

Safety and Handling

TSCA : No

Documents