missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylpyridinium Chloride, TCI America™

Manufacturer:  TCI America E054425G

 View more versions of this product

Catalog No. E054425G



Specifications

Specifications

1-Ethylpyridinium Chloride
25g
C7H10ClN
1-ethylpyridinium chloride, ethylpyridinium chloride, 1-ethylpyridin-1-ium chloride, unii-38bql5563g, pyridinium, 1-ethyl-, chloride, n-ethylpyridinium chloride, pyridinium, 1-ethyl-, chloride 1:1, 1-ethylpyridiniumchloride, pyridine ethochloride, ethylpyridiniumchloride
CC[N+]1=CC=CC=C1.[Cl-]
143.614
143.61
≥98.0% (T)
Blue-White
2294-38-4
MFCD03093293
AMFMJCAPWCXUEI-UHFFFAOYSA-M
1-ethylpyridin-1-ium;chloride
75299
Crystal-Lump at 20°C

Chemical Identifiers

2294-38-4
143.614
AMFMJCAPWCXUEI-UHFFFAOYSA-M
75299
CC[N+]1=CC=CC=C1.[Cl-]
C7H10ClN
MFCD03093293
1-ethylpyridinium chloride, ethylpyridinium chloride, 1-ethylpyridin-1-ium chloride, unii-38bql5563g, pyridinium, 1-ethyl-, chloride, n-ethylpyridinium chloride, pyridinium, 1-ethyl-, chloride 1:1, 1-ethylpyridiniumchloride, pyridine ethochloride, ethylpyridiniumchloride
1-ethylpyridin-1-ium;chloride
Safety and Handling

Safety and Handling

TSCA : Yes

Documents