missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylpyrazole, TCI America™

Manufacturer:  TCI America E103725ML

 View more versions of this product

Catalog No. E103725ML



Specifications

Specifications

1-Ethylpyrazole
Yellow
2817-71-2
MFCD03444367
FLNMQGISZVYIIK-UHFFFAOYSA-N
1-ethylpyrazole
498446
Clear Liquid at 20°C
137°C
25mL
C5H8N2
1993
CCN1C=CC=N1
96.133
96.13
≥98.0% (GC,T)

Chemical Identifiers

2817-71-2
96.133
FLNMQGISZVYIIK-UHFFFAOYSA-N
1-ethylpyrazole
C5H8N2
MFCD03444367
498446
CCN1C=CC=N1
Safety and Handling

Safety and Handling

TSCA : No

Documents