Learn More
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1629436
Description
In Room temperature phosphorescence of α-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application is described. The highest yields of 5-Bromopyrimidine (37%) were obtained by using 1-ethylpiperidine or triethylamine as condensing agent. Tributyltin hydride and 1-ethylpiperidine hypophosphite were mediated intermolecular radical additions to 2, 4, 6-trichlorophenyl vinyl sulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn Room temperature phosphorescence of α-bromonaphthalene induced by cyclodextrin in the presence of hexahydropyridine or 1-ethylpiperidine and its application is described. The highest yields of 5-Bromopyrimidine (37%) were obtained by using 1-ethylpiperidine or triethylamine as condensing agent. Tributyltin hydride and 1-ethylpiperidine hypophosphite were mediated intermolecular radical additions to 2, 4, 6-trichlorophenyl vinyl sulfonate.
Solubility
1-ethylpiperidine salt of benzylpenicillin is considerably less soluble in amyl acetate and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
1-Ethylpiperidine | |
-20°C | |
129°C to 131°C | |
C7H15N | |
MFCD00006507 | |
UN2386 | |
n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
1-ethylpiperidine | |
13007 | |
113.2 |
766-09-6 | |
0.825 | |
18°C (64°F) | |
1.444 | |
500 g | |
102643 | |
1-ethylpiperidine salt of benzylpenicillin is considerably less soluble in amyl acetate and acetone. | |
CCN1CCCCC1 | |
113.204 | |
CHEBI:39017 | |
99% |
Chemical Identifiers
766-09-6 | |
113.204 | |
HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
13007 | |
1-ethylpiperidine |
C7H15N | |
MFCD00006507 | |
n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
CHEBI:39017 | |
CCN1CCCCC1 |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P362-P370+P378q-P501c
H225-H301+H331-H312-H314-H318
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-ETHYLPIPERIDINE
EINECSNumber : 212-161-6
RTECSNumber : TN0250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only