missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethylnaphthalene, 98%

Manufacturer:  Alfa Aesar™ H5641306

 View more versions of this product

Catalog No. AAH5641306



Specifications

Specifications

1-Ethylnaphthalene
111°C (232°F)
Colorless to Yellow
5g
C12H12
1854417
ZMXIYERNXPIYFR-UHFFFAOYSA-N
1-ethylnaphthalene
14315
Liquid
1.008 g/mL
258°C to 260°C
-15°C
1127-76-0
MFCD00004049
.alpha.-ethylnaphthalene
CCC1=CC=CC2=CC=CC=C21
156.228
156.22
98%

Chemical Identifiers

1127-76-0
156.228
ZMXIYERNXPIYFR-UHFFFAOYSA-N
14315
CCC1=CC=CC2=CC=CC=C21
C12H12
MFCD00004049
.alpha.-ethylnaphthalene
1-ethylnaphthalene
Safety and Handling

Safety and Handling

EINECSNumber : 214-432-4

RTECSNumber : QJ6950000

TSCA : No

Recommended Storage : Ambient temperatures

Documents