Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethylimidazole-2-carboxaldehyde, 95%

Chemical Identifiers

Chemical Name or Material 1-Ethylimidazole-2-carboxaldehyde
CAS 111851-98-0
Formula Weight 124.14
MDL Number MFCD07380875
Beilstein 3538106
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6614303

 
alfa aesar™
H6614303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66143MD

 
alfa aesar™
H66143MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 1-Ethylimidazole-2-carboxaldehyde
CAS 111851-98-0
Assay 95%
Physical Form Solid
Beilstein 3538106
Quantity 1g
Storage Note 1 -30° to -10°C
MDL Number MFCD07380875
Safety and Handling

-30° to -10°C