missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylcyclopentanol, TCI America™

Manufacturer:  TCI America E078225G

 View more versions of this product

Catalog No. E078225G



Specifications

Specifications

1-Ethylcyclopentanol
Yellow
25g
C7H14O
1987
LPCWIFPJLFCXRS-UHFFFAOYSA-N
1-ethylcyclopentan-1-ol
137018
Clear Liquid at 20°C
153°C
-10°C
1462-96-0
MFCD00019281
1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, 1-ethylcyclopentanol, ksc181q7f, 1-ethyl-1-hydroxycyclopentane
CCC1(CCCC1)O
114.188
114.19
≥96.0% (GC)

Chemical Identifiers

1462-96-0
114.188
LPCWIFPJLFCXRS-UHFFFAOYSA-N
137018
CCC1(CCCC1)O
C7H14O
MFCD00019281
1-ethylcyclopentanol, cyclopentanol, 1-ethyl, 1-ethyl-1-cyclopentanol, acmc-1bzot, 1-ethylcyclopentanol, ksc181q7f, 1-ethyl-1-hydroxycyclopentane
1-ethylcyclopentan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents