missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethylcyclohexanol, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC453350050
Specifications
1-Ethylcyclohexanol | |
68°C | |
166.0°C | |
1940-18-7 | |
C8H16O | |
BUCJHJXFXUZJHL-UHFFFAOYSA-N | |
1-ethylcyclohexan-1-ol | |
16021 | |
97% |
0.9190g/mL | |
0.919 | |
5mL | |
96% min. (GC) | |
1-aethyl-cyclohexanol-1, 1-aethyl-cyclohexanol-1 german, 1-ethyl-1-cyclohexanol, 1-ethyl-cyclohexan-1-ol, 1-ethylcyclohexanol, 1ves5dbc5c, cyclohexanol, 1-ethyl, cyclohexanol, ethyl, ethylcyclohexanol, unii-1ves5dbc5c | |
CCC1(CCCCC1)O | |
128.22 | |
128.22 |
Chemical Identifiers
1940-18-7 | |
128.22 | |
1-aethyl-cyclohexanol-1, 1-aethyl-cyclohexanol-1 german, 1-ethyl-1-cyclohexanol, 1-ethyl-cyclohexan-1-ol, 1-ethylcyclohexanol, 1ves5dbc5c, cyclohexanol, 1-ethyl, cyclohexanol, ethyl, ethylcyclohexanol, unii-1ves5dbc5c | |
1-ethylcyclohexan-1-ol |
C8H16O | |
BUCJHJXFXUZJHL-UHFFFAOYSA-N | |
16021 | |
CCC1(CCCCC1)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS Signal Word: Warning