missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethylamino-2-propanol, TCI America™

Manufacturer:  TCI America E124425ML

 View more versions of this product

Catalog No. E124425ML



Specifications

Specifications

1-Ethylamino-2-propanol
Colorless
25mL
C5H13NO
2735
CCNCC(C)O
103.165
103.17
≥97.0% (T)
160°C
14°C
40171-86-6
MFCD00082193
UCYJVNBJCIZMTJ-UHFFFAOYSA-N
1-(ethylamino)propan-2-ol
3016164
Clear Liquid at 20°C

Chemical Identifiers

40171-86-6
103.165
UCYJVNBJCIZMTJ-UHFFFAOYSA-N
1-(ethylamino)propan-2-ol
C5H13NO
MFCD00082193
3016164
CCNCC(C)O
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1916

TSCA : No

Documents