Please login to your online account to display your discounted pricing

1-Ethyl-4-piperidone, 98%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  166181000

 View more versions of this product

Catalog No. AC166181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Formula Weight 127.19
Physical Form Viscous Liquid
Quantity 100g
Color Clear yellow to brown
EINECS Number 222-781-9
TSCA TSCA
Density 0.93g/mL
CAS Max % 100.0
Beilstein 21, V,6, 423
Chemical Name or Material 1-Ethyl-4-piperidone
Refractive Index 1.4620 to 1.4660 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H13NO
MDL Number MFCD00013356
CAS 3612-18-8
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Ethyl-4-piperidone
Name Note 98%
CAS 3612-18-8
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C7H13NO
Formula Weight 127.19
MDL Number MFCD00013356
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents