Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-piperidone, 98%

$60.97 - $719.04

Chemical Identifiers

Chemical Name or Material 1-Ethyl-4-piperidone
CAS 3612-18-8
Molecular Formula C7H13NO
Formula Weight 127.19g/mol
MDL Number MFCD00013356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB212690B

 
alfa aesar™
B21269-0B
1000g Each for $719.04

AAB2126918

 
alfa aesar™
B21269-18
50g Each for $60.97

AAB2126930

 
alfa aesar™
B21269-30
250g Each for $218.84
Description & Specifications

Specifications

Boiling Point 46° to 48°C (1mmHg)
Quantity 1000g
Flash Point 68°C (156°F)
Chemical Name or Material 1-Ethyl-4-piperidone
CAS 3612-18-8
Assay 98%
Molecular Formula C7H13NO
Formula Weight 127.19g/mol
MDL Number MFCD00013356
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used to produce 1-ethyl-1-methyl-4-oxo-piperidinium; iodide when reacted with iodomethane and also in the preparation of 3,5-bisarylmethylene-1-ethyl-4-piperidones. Piperidones exhibit anticancer, anti-inflammatory, and antimicrobial activity, and also inhibit the NF-κB signaling pathway. 4-piperidone are important intermediates in alkaloid synthesis and in the preparation of medicinal agents. Mannich-Michael, and aza-Diels-Alder reactions involving an imino dienophile and a conjugated diene and enones have been applied to the synthesis of piperidine derivatives.

Solubility
Miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.