missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-4-methylpyridinium Bromide, TCI America™

Manufacturer:  TCI America E127225G

 View more versions of this product

Catalog No. E127225G



Specifications

Specifications

1-Ethyl-4-methylpyridinium Bromide
129°C
32353-49-4
MFCD18384825
CC[N+]1=CC=C(C=C1)C.[Br-]
202.095
202.10
≥98.0% (HPLC,T)
Undesignated
25g
C8H12BrN
YZBRZOWPIBQUEJ-UHFFFAOYSA-M
1-ethyl-4-methylpyridin-1-ium;bromide
14358228
Crystal-Powder at 20°C

Chemical Identifiers

32353-49-4
202.095
YZBRZOWPIBQUEJ-UHFFFAOYSA-M
1-ethyl-4-methylpyridin-1-ium;bromide
C8H12BrN
MFCD18384825
14358228
CC[N+]1=CC=C(C=C1)C.[Br-]
Safety and Handling

Safety and Handling

TSCA : No

Documents