Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-fluorobenzene, 97%

Chemical Identifiers

Chemical Name or Material 1-Ethyl-4-fluorobenzene
CAS 459-47-2
Molecular Formula C8H9F
Formula Weight 124.16
MDL Number MFCD00039218
UN Number UN1993
Beilstein 1856987
Synonym 4-Fluoroethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0594006

  SDS
alfa aesar™
L0594006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0594014

  SDS
alfa aesar™
L0594014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.16
Chemical Name or Material 1-Ethyl-4-fluorobenzene
Assay 97%
Molecular Formula C8H9F
MDL Number MFCD00039218
UN Number UN1993
Beilstein 1856987
Boiling Point 139°C
Health Hazard 1 Warning
Quantity 5g
Flash Point 30°C (86°F)
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
CAS 459-47-2
Synonym 4-Fluoroethylbenzene

Applications
1-Ethyl-4-fluorobenzene is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.