Please login to your online account to display your discounted pricing

1-Ethyl-4-eth-1-ynylbenzene, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB09906DA

 View more versions of this product

Catalog No. BTB09906DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 130.19
Quantity 1g
Assay 97%
Density 0.93g/mL
CAS Max % 100.0
Assay Percent Range 97%
Chemical Name or Material 1-Ethyl-4-eth-1-ynylbenzene
Linear Formula HC2CC6H4CH2CH3
MDL Number MFCD00173887
CAS Min % 96.5
Packaging Amber glass bottle
Molecular Formula C10H10
CAS 40307-11-7
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Ethyl-4-eth-1-ynylbenzene
CAS 40307-11-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10
Linear Formula HC2CC6H4CH2CH3
Formula Weight 130.19
MDL Number MFCD00173887
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents