missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-(4-nitrophenyl)piperazine, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 115619-00-6
Molecular Formula C12H17N3O2
Molecular Weight (g/mol) 235.287
MDL Number MFCD00614372
InChI Key XEWICNRVCQLKIG-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-nitrophenyl piperazine, 1-ethyl-4-4-nitro-phenyl-piperazine, piperazine,1-ethyl-4-4-nitrophenyl, piperazine, 1-ethyl-4-4-nitrophenyl, acmc-1bqhi, cbmicro_027169, 4-ethyl-1-4-nitrophenyl piperazine
PubChem CID 2857549
SMILES CCN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63555MD View Documents Alfa Aesar™
H63555MD
250mg Each for $28.84
 
AAH6355503 View Documents Alfa Aesar™
H6355503
1g Each for $85.14
 
AAH6355506 View Documents Alfa Aesar™
H6355506
5g Each for $337.73
 
Specifications

Chemical Identifiers

115619-00-6
235.287
XEWICNRVCQLKIG-UHFFFAOYSA-N
2857549
C12H17N3O2
MFCD00614372
1-ethyl-4-4-nitrophenyl piperazine, 1-ethyl-4-4-nitro-phenyl-piperazine, piperazine,1-ethyl-4-4-nitrophenyl, piperazine, 1-ethyl-4-4-nitrophenyl, acmc-1bqhi, cbmicro_027169, 4-ethyl-1-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

250mg
C12H17N3O2
1-ethyl-4-4-nitrophenyl piperazine, 1-ethyl-4-4-nitro-phenyl-piperazine, piperazine,1-ethyl-4-4-nitrophenyl, piperazine, 1-ethyl-4-4-nitrophenyl, acmc-1bqhi, cbmicro_027169, 4-ethyl-1-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
2857549
Solid
1-Ethyl-4-(4-nitrophenyl)piperazine
115619-00-6
MFCD00614372
XEWICNRVCQLKIG-UHFFFAOYSA-N
235.287
235.28
97%