Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene, 99+%

$79.58 - $418.69

Chemical Identifiers

Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
CAS 63221-88-5
Molecular Formula C17H16O
Formula Weight 236.31
MDL Number MFCD00807428
Synonym 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2703809

  SDS
alfa aesar™
H2703809
10g Each for $418.69

AAH2703803

  SDS
alfa aesar™
H2703803
1g Each for $79.58
Description & Specifications

Specifications

Formula Weight 236.31
Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
Molecular Formula C17H16O
CAS 63221-88-5
Boiling Point 151° to 153°C (1mmHg)
Assay ≥99%
MDL Number MFCD00807428
Synonym 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene
Melting Point 35°C (N)
Quantity 10g