missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-(4-fluoro-2-nitrophenyl)piperazine, 97%

Chemical Identifiers

CAS 1285896-76-5
Molecular Formula C12H16FN3O2
Molecular Weight (g/mol) 253.277
InChI Key IWKXGRRMWASTNC-UHFFFAOYSA-N
Synonym 1-ethyl-4-4-fluoro-2-nitrophenyl piperazine
PubChem CID 52438092
IUPAC Name 1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
SMILES CCN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63999MD View Documents Alfa Aesar™
H63999MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6399903 View Documents Alfa Aesar™
H6399903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6399906 View Documents Alfa Aesar™
H6399906
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1285896-76-5
253.277
1-ethyl-4-4-fluoro-2-nitrophenyl piperazine
1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
C12H16FN3O2
IWKXGRRMWASTNC-UHFFFAOYSA-N
52438092
CCN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

250mg
C12H16FN3O2
IWKXGRRMWASTNC-UHFFFAOYSA-N
1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
52438092
Solid
1-Ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
1285896-76-5
1-ethyl-4-4-fluoro-2-nitrophenyl piperazine
CCN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]
253.277
253.27
97%