missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine, 97%

$35.93 - $422.54

Chemical Identifiers

CAS 694501-34-3
Molecular Formula C12H16FN3O2
Molecular Weight (g/mol) 253.277
InChI Key XMJXGMZBNFICRS-UHFFFAOYSA-N
Synonym 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl, 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine
PubChem CID 2947839
SMILES CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63843MD View Documents Alfa Aesar™
H63843MD
250mg Each for $35.93
 
AAH6384303 View Documents Alfa Aesar™
H6384303
1g Each for $106.26
 
AAH6384306 View Documents Alfa Aesar™
H6384306
5g Each for $422.54
 
Specifications

Chemical Identifiers

694501-34-3
253.277
1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl, 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
C12H16FN3O2
XMJXGMZBNFICRS-UHFFFAOYSA-N
2947839

Specifications

250mg
C12H16FN3O2
1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl, 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
2947839
Solid
1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine
694501-34-3
15470780
XMJXGMZBNFICRS-UHFFFAOYSA-N
253.277
253.27
97%