missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6384303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine | |
C12H16FN3O2 | |
15470780 | |
XMJXGMZBNFICRS-UHFFFAOYSA-N | |
253.277 | |
253.27 | |
Solid |
694501-34-3 | |
1 g | |
1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl | |
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F | |
2947839 | |
97% |
Chemical Identifiers
694501-34-3 | |
253.277 | |
2947839 |
C12H16FN3O2 | |
XMJXGMZBNFICRS-UHFFFAOYSA-N | |
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures