Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate, 98+%

$83.93 - $307.38

Chemical Identifiers

Chemical Name or Material 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate
CAS 145022-44-2
Molecular Formula C7H11F3N2O3S
Formula Weight 260.24
MDL Number MFCD00210008
Synonym 1-Ethyl-3-methylimidazolium triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1976404

  SDS
alfa aesar™
L1976404
2g Each for $83.93

AAL1976409

  SDS
alfa aesar™
L1976409
10g Each for $307.38
Description & Specifications

Specifications

Formula Weight 260.24
Chemical Name or Material 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate
Molecular Formula C7H11F3N2O3S
MDL Number MFCD00210008
CAS 145022-44-2
Synonym 1-Ethyl-3-methylimidazolium triflate
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay ≥98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 2g

Applications
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.