missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Tricyanomethanide 98.0+%, TCI America™

$195.76 - $587.29

Chemical Identifiers

CAS 666823-18-3
Molecular Formula C10H11N5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E12985G
View Documents Promotion Details
TCI America
E12985G
5 g
Each for $195.76
 
E129825G
View Documents Promotion Details
TCI America
E129825G
25 g
Each for $587.29
 
Description

Description

Specifications

Chemical Identifiers

666823-18-3
C10H11N5

Specifications

666823-18-3
C10H11N5
201.23
Liquid
-10°C
5 g
≥98.0% (HPLC,N)
1-Ethyl-3-methylimidazolium Tricyanomethanide
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'