missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Thiocyanate, TCI America™

Manufacturer:  TCI America E077625G

 View more versions of this product

Catalog No. E077625G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Thiocyanate
-6°C
331717-63-6
MFCD06798182
VASPYXGQVWPGAB-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;thiocyanate
16211115
Clear Liquid at 20°C
Undesignated
25g
C7H11N3S
1-ethyl-3-methylimidazolium thiocyanate, basionics® vs 01, dsstox_cid_27860, dsstox_rid_82613, dsstox_gsid_47884, 1-methyl-3-ethylimidazolium thiocyanate, 1-ethyl-3-methyl imidazolium thio-cyanate, 1-ethyl-3-methyl-1h-imidazol-3-ium thiocyanate, 1-ethyl-3-methylimidazolium thiocyanate h-nmr, hplc, 1-ethyl-3-methylimidazolium thiocyanate, for electrochemistry t
CCN1C=C[N+](=C1)C.C(#N)[S-]
169.246
169.25
≥98.0% (HPLC,T)

Chemical Identifiers

331717-63-6
169.246
VASPYXGQVWPGAB-UHFFFAOYSA-M
16211115
CCN1C=C[N+](=C1)C.C(#N)[S-]
C7H11N3S
MFCD06798182
1-ethyl-3-methylimidazolium thiocyanate, basionics® vs 01, dsstox_cid_27860, dsstox_rid_82613, dsstox_gsid_47884, 1-methyl-3-ethylimidazolium thiocyanate, 1-ethyl-3-methyl imidazolium thio-cyanate, 1-ethyl-3-methyl-1h-imidazol-3-ium thiocyanate, 1-ethyl-3-methylimidazolium thiocyanate h-nmr, hplc, 1-ethyl-3-methylimidazolium thiocyanate, for electrochemistry t
1-ethyl-3-methylimidazol-3-ium;thiocyanate
Safety and Handling

Safety and Handling

TSCA : No

Documents