missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Methyl Sulfate, TCI America™

Manufacturer:  TCI America E130825G

 View more versions of this product

Catalog No. E130825G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Methyl Sulfate
25g
C7H14N2O4S
1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc
CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-]
222.259
222.26
≥98.0% (HPLC)
Undesignated
516474-01-4
MFCD08457697
BXSDLSWVIAITRQ-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;methyl sulfate
16211409
Clear Liquid at 20°C

Chemical Identifiers

516474-01-4
222.259
BXSDLSWVIAITRQ-UHFFFAOYSA-M
16211409
CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-]
C7H14N2O4S
MFCD08457697
1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc
1-ethyl-3-methylimidazol-3-ium;methyl sulfate
Safety and Handling

Safety and Handling

TSCA : No

Documents