missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Methanesulfonate, TCI America™

Manufacturer:  TCI America E075525G

 View more versions of this product

Catalog No. E075525G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Methanesulfonate
25g
C7H14N2O3S
3265
IXLWEDFOKSJYBD-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;methanesulfonate
16211849
Clear Liquid at 20°C
Undesignated
145022-45-3
MFCD06798171
1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics® st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate
CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]
206.26
206.26
≥98.0% (N)

Chemical Identifiers

145022-45-3
206.26
IXLWEDFOKSJYBD-UHFFFAOYSA-M
16211849
CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]
C7H14N2O3S
MFCD06798171
1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics® st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate
1-ethyl-3-methylimidazol-3-ium;methanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents