missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Ethyl-3-methylimidazolium Methanesulfonate 98.0+%, TCI America™
$51.43 - $142.67
Chemical Identifiers
CAS | 145022-45-3 |
---|---|
Molecular Formula | C7H14N2O3S |
Molecular Weight (g/mol) | 206.26 |
MDL Number | MFCD06798171 |
InChI Key | IXLWEDFOKSJYBD-UHFFFAOYSA-M |
Synonym | 1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics™ st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate |
PubChem CID | 16211849 |
IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;methanesulfonate |
SMILES | CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-] |
Chemical Identifiers
145022-45-3 | |
206.26 | |
IXLWEDFOKSJYBD-UHFFFAOYSA-M | |
16211849 | |
CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-] |
C7H14N2O3S | |
MFCD06798171 | |
1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics™ st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate | |
1-ethyl-3-methylimidazol-3-ium;methanesulfonate |
Specifications
145022-45-3 | |
MFCD06798171 | |
3265 | |
IXLWEDFOKSJYBD-UHFFFAOYSA-M | |
1-ethyl-3-methylimidazol-3-ium;methanesulfonate | |
16211849 | |
≥98.0% (N) | |
1-Ethyl-3-methylimidazolium Methanesulfonate |
C7H14N2O3S | |
5 g | |
1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics™ st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate | |
CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-] | |
206.26 | |
206.26 | |
Liquid |
Safety and Handling
TSCA : No