Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-3-methylimidazolium methanesulfonate, 99%

$40.42 - $40.42

Chemical Identifiers

Chemical Name or Material 1-Ethyl-3-methylimidazolium methanesulfonate
CAS 145022-45-3
Molecular Formula C7H14N2O3S
Formula Weight 206.26g/mol
MDL Number MFCD06798171
UN Number UN3265
Synonym ECOENG 110
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2691309

 
alfa aesar™
H26913-09
10g Each for $40.42
Description & Specifications

Specifications

Melting Point 35°C
Quantity 10g
Flash Point 66°C (152°F)
Chemical Name or Material 1-Ethyl-3-methylimidazolium methanesulfonate
CAS 145022-45-3
Assay 99%
Molecular Formula C7H14N2O3S
Formula Weight 206.26g/mol
MDL Number MFCD06798171
UN Number UN3265
Synonym ECOENG 110
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Applications
Used as an electrolyte for the electrooxidative polymerization of pyrrole. Used for electroplating of aluminum onto steel, electroplating of chromium,„ electropolishing of steel and other metal surfaces. In polymer and biopolymer processing used as„ solvent for regenerating cellulose,„ enabling technology for manufacturing cellulose, derivatives„ dissolution and separation of lignocellulosic biomas.

Solubility
Soluble in water. Soluble in acetone, methanol .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.