missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-3-methylimidazolium iodide, 97%

Manufacturer:  Alfa Aesar™ H5940418

 View more versions of this product

Catalog No. AAH5940418



Description

Description

Applications
It is used as OLED materials.

Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.

Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Ethyl-3-methylimidazolium iodide
White to Yellow
35935-34-3
MFCD03701101
1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt, 1-ethyl-3-methylimidazolium iodide
CCN1C=C[N+](=C1)C.[I-]
238.072
238.07
97%
Moisture Sensitive
50g
C6H11IN2
5160333
IKQCDTXBZKMPBB-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;iodide
11075478
Powder

Chemical Identifiers

35935-34-3
238.072
IKQCDTXBZKMPBB-UHFFFAOYSA-M
11075478
CCN1C=C[N+](=C1)C.[I-]
C6H11IN2
MFCD03701101
1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt, 1-ethyl-3-methylimidazolium iodide
1-ethyl-3-methylimidazol-3-ium;iodide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents