missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Hydrogen Sulfate, TCI America™

Manufacturer:  TCI America E07545G

 View more versions of this product

Catalog No. E07545G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Hydrogen Sulfate
5g
C6H12N2O4S
3265
HZKDSQCZNUUQIF-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;hydrogen sulfate
16217215
Clear Liquid at 20°C
Yellow
412009-61-1
MFCD06798195
1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics® ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1
CCN1C=C[N+](=C1)C.OS(=O)(=O)[O-]
208.232
208.23
≥98.0% (N)

Chemical Identifiers

412009-61-1
208.232
HZKDSQCZNUUQIF-UHFFFAOYSA-M
16217215
CCN1C=C[N+](=C1)C.OS(=O)(=O)[O-]
C6H12N2O4S
MFCD06798195
1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics® ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1
1-ethyl-3-methylimidazol-3-ium;hydrogen sulfate
Safety and Handling

Safety and Handling

TSCA : No

Documents