missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Ethyl Sulfate, TCI America™

Manufacturer:  TCI America E06505G

 View more versions of this product

Catalog No. E06505G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Ethyl Sulfate
5g
C8H16N2O4S
VRFOKYHDLYBVAL-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
12095229
Clear Liquid at 20°C
Yellow
342573-75-5
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics® lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]
236.286
236.29
≥98.0% (N)

Chemical Identifiers

342573-75-5
236.286
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics® lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate
C8H16N2O4S
VRFOKYHDLYBVAL-UHFFFAOYSA-M
12095229
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents