missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-methylimidazolium Chloride [for Molten Salt], TCI America™

Manufacturer:  TCI America E049025G

 View more versions of this product

Catalog No. E049025G



Specifications

Specifications

1-Ethyl-3-methylimidazolium Chloride [for Molten Salt]
84°C
65039-09-0
MFCD00074843
BMQZYMYBQZGEEY-UHFFFAOYSA-M
1-ethyl-3-methylimidazol-3-ium;chloride
2734160
146.62
≥98.0% (HPLC,T)
Undesignated
25g
C6H11ClN2
1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics® st 80, dsstox_cid_21180
CCN1C=C[N+](=C1)C.[Cl-]
146.618
CHEBI:61327
Crystal-Powder at 20°C

Chemical Identifiers

65039-09-0
146.618
BMQZYMYBQZGEEY-UHFFFAOYSA-M
2734160
1-ethyl-3-methylimidazol-3-ium;chloride
C6H11ClN2
MFCD00074843
1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics® st 80, dsstox_cid_21180
CHEBI:61327
CCN1C=C[N+](=C1)C.[Cl-]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents