missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-3-methyl-4-piperidone, 98%

$41.32 - $437.96

Chemical Identifiers

CAS 3612-16-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD01318144
InChI Key DZPMPRBBBQMILW-UHFFFAOYSA-N
Synonym 1-ethyl-3-methyl-4-piperidone, acmc-20aova, 1-ethyl-3-methyl-piperidin-4-one
PubChem CID 2733442
IUPAC Name 1-ethyl-3-methylpiperidin-4-one
SMILES CCN1CCC(=O)C(C1)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1712403 View Documents Alfa Aesar™
L1712403
1g Each for $41.32
 
AAL1712406 View Documents Alfa Aesar™
L1712406
5g Each for $118.43
 
AAL1712414 View Documents Alfa Aesar™
L1712414
25g Each for $437.96
 
Specifications

Chemical Identifiers

3612-16-6
141.214
DZPMPRBBBQMILW-UHFFFAOYSA-N
2733442
CCN1CCC(=O)C(C1)C
C8H15NO
MFCD01318144
1-ethyl-3-methyl-4-piperidone, acmc-20aova, 1-ethyl-3-methyl-piperidin-4-one
1-ethyl-3-methylpiperidin-4-one

Specifications

72°C (7.6mmHg)
3612-16-6
MFCD01318144
1-ethyl-3-methyl-4-piperidone, acmc-20aova, 1-ethyl-3-methyl-piperidin-4-one
CCN1CCC(=O)C(C1)C
141.214
141.21
1-Ethyl-3-methyl-4-piperidone
1g
C8H15NO
1524485
DZPMPRBBBQMILW-UHFFFAOYSA-N
1-ethyl-3-methylpiperidin-4-one
2733442
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning