Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid, 97%

$78.35 - $280.15

Chemical Identifiers

Chemical Name or Material 1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
CAS 50920-65-5
Molecular Formula C7H10N2O2
Formula Weight 154.17
MDL Number MFCD00156147
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1156103

 
alfa aesar™
A1156103
1g Each for $78.35

AAA1156106

 
alfa aesar™
A1156106
5g Each for $280.15
Description & Specifications

Specifications

Formula Weight 154.17
Chemical Name or Material 1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Assay 97%
Molecular Formula C7H10N2O2
MDL Number MFCD00156147
CAS 50920-65-5
Melting Point 137° to 140°C
Quantity 1g