Please login to your online account to display your discounted pricing

1-Ethyl-3-hydroxypiperidine ≥98%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  118580050

 View more versions of this product

Catalog No. AC118580050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 98.0 to 101.0% total base (on anhydrous substance), ≥98.0% (GC)
Physical Form Viscous Liquid
Formula Weight 129.2
Quantity 5g
Color Clear
EINECS Number 236-584-0
TSCA TSCA
Density 0.97g/mL
Merck Index 15, 3897
CAS Min % 98.0
CAS Max % 100.0
Refractive Index 1.4754 to 1.4774 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Water 0.5% max. (K.F.)
Synonym 1-Ethyl-3-piperidinol
Name Note 98+%
Molecular Formula C7H15NO
Chemical Name or Material 1-Ethyl-3-hydroxypiperidine
Assay Percent Range 98+%
MDL Number MFCD00006508
CAS 13444-24-1

Specifications

Chemical Name or Material 1-Ethyl-3-hydroxypiperidine
Name Note 98+%
CAS 13444-24-1
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0 to 101.0% total base (on anhydrous substance), ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00006508
Synonym 1-Ethyl-3-piperidinol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents