missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-3-hydroxypiperidine ≥98%, Acros Organics

$36.41 - $36.41

Chemical Identifiers

CAS 13444-24-1
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00006508
Synonym 1-Ethyl-3-piperidinol
Chemical Name or Material 1-Ethyl-3-hydroxypiperidine, 98+%
Assay 98.0 to 101.0% total base (on anhydrous substance), ≥98.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC118580050 SDS Thermo Fisher Scientific
118580050
5g Each for $36.41
Specifications Description & Specifications

Chemical Identifiers

13444-24-1
100.0
C7H15NO
MFCD00006508
1-Ethyl-3-hydroxypiperidine, 98+%
98.0
98+%
129.2
1-Ethyl-3-piperidinol
98.0 to 101.0% total base (on anhydrous substance), ≥98.0% (GC)

Specifications

13444-24-1
100.0
Authentic
98+%
1.4754 to 1.4774 (20°C, 589nm)
MFCD00006508
15, 3897
Clear
1-Ethyl-3-hydroxypiperidine, 98+%
98.0
0.97g/mL
Viscous Liquid
C7H15NO
129.2
0.5% max. (K.F.)
1-Ethyl-3-piperidinol
5g
98.0 to 101.0% total base (on anhydrous substance), ≥98.0% (GC)

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-584-0

TSCA : TSCA