missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-2-pyrrolidone, TCI America™

Manufacturer:  TCI America E0358250ML

 View more versions of this product

Catalog No. E0358250ML



Specifications

Specifications

1-Ethyl-2-pyrrolidone
Yellow
2687-91-4
MFCD00003199
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethylpyrrolidin-2-one
17595
Clear Liquid at 20°C
218°C
250mL
C6H11NO
1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone
CCN1CCCC1=O
113.16
113.16
≥98.0% (GC)

Chemical Identifiers

2687-91-4
113.16
ZFPGARUNNKGOBB-UHFFFAOYSA-N
17595
CCN1CCCC1=O
C6H11NO
MFCD00003199
1-ethyl-2-pyrrolidinone, n-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone
1-ethylpyrrolidin-2-one
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0113

RTECSNumber : UY5769250

TSCA : Yes

Documents