Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-2-phenylindole, 98%

$20.04 - $71.12

Chemical Identifiers

Chemical Name or Material 1-Ethyl-2-phenylindole
CAS 13228-39-2
Molecular Formula C16H15N
Formula Weight 221.3
MDL Number MFCD00031481
Beilstein 168581
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1122809

  SDS
alfa aesar™
L1122809
10g Each for $20.04

AAL1122818

  SDS
alfa aesar™
L1122818
50g Each for $71.12
Description & Specifications

Specifications

Formula Weight 221.3
MDL Number MFCD00031481
CAS 13228-39-2
Quantity 10g
Melting Point 85° to 86°C
Beilstein 168581
Molecular Formula C16H15N
Chemical Name or Material 1-Ethyl-2-phenylindole
Assay 98%