missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1058414

 View more versions of this product

Catalog No. AAA1058414



Description

Description

1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

Solubility
Soluble in Water.

Notes
Precursor in synthesis of carbocyanine dyes.
Specifications

Specifications

1-Ethyl-2-methylquinolinium iodide
231°C (decomposition)
MFCD00031829
Light sensitive
Soluble in Water.
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]
299.155
299.16
606-55-3
C12H14IN
25 g
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
OEVSHJVOKFWBJY-UHFFFAOYSA-M
1-ethyl-2-methylquinolin-1-ium;iodide
69076
97%

Chemical Identifiers

606-55-3
299.155
OEVSHJVOKFWBJY-UHFFFAOYSA-M
69076
CC[N+]1=C(C=CC2=CC=CC=C21)C.[I-]
C12H14IN
MFCD00031829
1-ethyl-2-methylquinolinium iodide, quinaldine ethiodide, 1-ethylquinaldinium iodide, n-ethylquinaldinium iodide, 2-methylquinoline ethiodide, quinaldinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-2-methyl-, iodide, 2-methyl-1-ethylquinolinium iodide, 1-ethyl-2-methylquinolin-1-ium iodide, quinolinium, 1-ethyl-2-methyl-, iodide 1:1
1-ethyl-2-methylquinolin-1-ium;iodide
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-120-7

RTECSNumber : VA0740000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only