Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 98%

Chemical Identifiers

Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate
CAS 174899-72-0
Formula Weight 274.26
MDL Number MFCD07784438
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5972606

 
alfa aesar™
H5972606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5972618

 
alfa aesar™
H5972618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.26
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate
Physical Form Liquid
Assay 98%
MDL Number MFCD07784438
CAS 174899-72-0
Sensitivity Moisture Sensitive
Color Pale yellow
Quantity 5g