missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate, 98%

$76.19 - $306.88

Chemical Identifiers

CAS 292140-86-4
Molecular Formula C7H13F6N2P
Molecular Weight (g/mol) 270.159
MDL Number MFCD08458503
InChI Key XLKYEJKYOZNSDQ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
PubChem CID 50878556
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6022809 View Documents Alfa Aesar™
H6022809
10g Each for $76.19
 
AAH6022818 View Documents Alfa Aesar™
H6022818
50g Each for $306.88
 
Specifications

Chemical Identifiers

292140-86-4
270.159
XLKYEJKYOZNSDQ-UHFFFAOYSA-N
50878556
CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F
C7H13F6N2P
MFCD08458503
1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
1-ethyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate

Specifications

10g
C7H13F6N2P
1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F
270.159
270.16
1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate
292140-86-4
MFCD08458503
XLKYEJKYOZNSDQ-UHFFFAOYSA-N
1-ethyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
50878556
98%