missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-1-(o-tolyl)hydrazine, TCI America™

Manufacturer:  TCI America E055225G

 View more versions of this product

Catalog No. E055225G



Specifications

Specifications

1-Ethyl-1-(o-tolyl)hydrazine
25g
C9H14N2
FGGBNBLMSDRPEA-UHFFFAOYSA-N
1-ethyl-1-(2-methylphenyl)hydrazine
15591717
Clear Liquid at 20°C
Yellow
910041-10-0
MFCD03844791
CCN(C1=CC=CC=C1C)N
150.225
150.23
≥98.0% (T)

Chemical Identifiers

910041-10-0
150.225
FGGBNBLMSDRPEA-UHFFFAOYSA-N
1-ethyl-1-(2-methylphenyl)hydrazine
C9H14N2
MFCD03844791
15591717
CCN(C1=CC=CC=C1C)N
Safety and Handling

Safety and Handling

TSCA : No

Documents