missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethyl-1-(m-tolyl)hydrazine, TCI America™

Manufacturer:  TCI America E055325G

 View more versions of this product

Catalog No. E055325G



Specifications

Specifications

1-Ethyl-1-(m-tolyl)hydrazine
25g
C9H14N2
KSNDNKBQRCZUFZ-UHFFFAOYSA-N
1-ethyl-1-(3-methylphenyl)hydrazine
44630285
Clear Liquid at 20°C
Yellow
99717-68-7
MFCD03844792
CCN(C1=CC=CC(=C1)C)N
150.225
150.23
≥98.0% (T)

Chemical Identifiers

99717-68-7
150.225
KSNDNKBQRCZUFZ-UHFFFAOYSA-N
1-ethyl-1-(3-methylphenyl)hydrazine
C9H14N2
MFCD03844792
44630285
CCN(C1=CC=CC(=C1)C)N
Safety and Handling

Safety and Handling

TSCA : No

Documents