Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethoxymethyl-2-iodoimidazole, 97%

$85.43 - $253.14

Chemical Identifiers

Chemical Name or Material 1-Ethoxymethyl-2-iodoimidazole
CAS 146697-87-2
Molecular Formula C6H9IN2O
Formula Weight 252.05
MDL Number MFCD03788568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5186503

  SDS
alfa aesar™
H5186503
1g Each for $253.14

AAH51865MD

  SDS
alfa aesar™
H51865MD
250mg Each for $85.43
Description & Specifications

Specifications

Formula Weight 252.05
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Chemical Name or Material 1-Ethoxymethyl-2-iodoimidazole
MDL Number MFCD03788568
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
CAS 146697-87-2
Molecular Formula C6H9IN2O
Assay 97%
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.