Learn More
(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 170290250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(1-Ethoxyethylidene)malononitrile | |
5417-82-3 | |
100.0 | |
99% min. (GC) | |
C7H8N2O | |
MFCD00001852 | |
1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene propanedinitrile, propanedinitrile, 1-ethoxyethylidene, 1-ethoxyethylidenemalononitrile, 1-ethoxyethylidene propanedinitrile, 2-1-ethoxyethylidene-propanedinitrile, propanedinitrile, 2-1-ethoxyethylidene, ethoxyethylidene methane-1,1-dicarbonitrile, malononitrile, 1-ethoxyethylidene-6ci,7ci,8ci | |
CCOC(=C(C#N)C#N)C | |
136.15 | |
136.15 |
99+% | |
99.0 | |
Authentic | |
Plastic bottle | |
CH3C(OC2H5)=C(CN)2 | |
25g | |
BOSVWXDDFBSSIZ-UHFFFAOYSA-N | |
2-(1-ethoxyethylidene)propanedinitrile | |
79450 | |
99+% |
Chemical Identifiers
5417-82-3 | |
136.15 | |
BOSVWXDDFBSSIZ-UHFFFAOYSA-N | |
79450 | |
CCOC(=C(C#N)C#N)C |
C7H8N2O | |
MFCD00001852 | |
1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene propanedinitrile, propanedinitrile, 1-ethoxyethylidene, 1-ethoxyethylidenemalononitrile, 1-ethoxyethylidene propanedinitrile, 2-1-ethoxyethylidene-propanedinitrile, propanedinitrile, 2-1-ethoxyethylidene, ethoxyethylidene methane-1,1-dicarbonitrile, malononitrile, 1-ethoxyethylidene-6ci,7ci,8ci | |
2-(1-ethoxyethylidene)propanedinitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENT
GHS Signal Word: Warning
EINECSNumber : 226-521-5
RTECSNumber : TY1510000
TSCA : TSCA
RUO â Research Use Only