Please login to your online account to display your discounted pricing

(1-Ethoxycyclopropoxy)trimethylsilane, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368901000

 View more versions of this product

Catalog No. AC368901000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 174.52
Quantity 100g
Color Clear colorless
Density 0.867g/mL
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H18O2Si
Linear Formula CH5OC3H4OSi(CH3)3
CAS 27374-25-0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
CAS Min % 98.5
Infrared Spectrum Authentic
Refractive Index 1.4070 to 1.4090 (20°C, 589nm)
Packaging Glass bottle
Synonym Cyclopropanone ethyletrimethylsilyl acetal
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00074986
Chemical Name or Material (1-Ethoxycyclopropoxy)trimethylsilane
Name Note 99%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (1-Ethoxycyclopropoxy)trimethylsilane
Name Note 99%
CAS 27374-25-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H18O2Si
Linear Formula CH5OC3H4OSi(CH3)3
Formula Weight 174.52
MDL Number MFCD00074986
Synonym Cyclopropanone ethyletrimethylsilyl acetal
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents