missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethoxycarbonyl-4-piperidone, TCI America™

Manufacturer:  TCI America E0403500G

 View more versions of this product

Catalog No. E0403500G



Specifications

Specifications

1-Ethoxycarbonyl-4-piperidone
Yellow
29976-53-2
MFCD00006188
LUBGFMZTGFXIIN-UHFFFAOYSA-N
ethyl 4-oxopiperidine-1-carboxylate
34767
Clear Liquid at 20°C
151°C
500g
C8H13NO3
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
CCOC(=O)N1CCC(=O)CC1
171.196
171.20
≥97.0% (GC)

Chemical Identifiers

29976-53-2
171.196
LUBGFMZTGFXIIN-UHFFFAOYSA-N
34767
CCOC(=O)N1CCC(=O)CC1
C8H13NO3
MFCD00006188
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
ethyl 4-oxopiperidine-1-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents