Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethoxycarbonyl-4-piperidone, 99%

$51.11 - $741.93

Chemical Identifiers

Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone
CAS 29976-53-2
Molecular Formula C8H13NO3
Formula Weight 171.2
MDL Number MFCD00006188
Beilstein 640967
Synonym N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1183309

  SDS
alfa aesar™
A1183309
10g Each for $51.11

AAA1183318

  SDS
alfa aesar™
A1183318
50g Each for $181.64

AAA1183330

  SDS
alfa aesar™
A1183330
250g Each for $741.93
Description & Specifications

Specifications

Formula Weight 171.2
Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone
Assay 99%
Molecular Formula C8H13NO3
MDL Number MFCD00006188
CAS 29976-53-2
Beilstein 640967
Synonym N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 87°C (188°F)
Boiling Point 95° to 98°C (1mmHg)
Quantity 10g

Applications
1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

Solubility
Soluble in water.500 g/L at 20°C

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Protect from heat. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.