missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethoxycarbonyl-4-piperidone, 99%

Manufacturer:  Alfa Aesar™ A1183330

 View more versions of this product

Catalog No. AAA1183330



Description

Description

Applications
1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

Solubility
Soluble in water.500 g/L at 20°C

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Protect from heat. Incompatible with oxidizing agents.
Specifications

Specifications

1-Ethoxycarbonyl-4-piperidone
95°C to 98°C (1mmHg)
29976-53-2
C8H13NO3
640967
LUBGFMZTGFXIIN-UHFFFAOYSA-N
ethyl 4-oxopiperidine-1-carboxylate
34767
87°C (188°F)
250g
99%
MFCD00006188
N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
CCOC(=O)N1CCC(=O)CC1
171.196
171.2

Chemical Identifiers

29976-53-2
171.196
LUBGFMZTGFXIIN-UHFFFAOYSA-N
34767
CCOC(=O)N1CCC(=O)CC1
C8H13NO3
MFCD00006188
N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
ethyl 4-oxopiperidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents