missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethoxy-4-ethynylbenzene, TCI America™

Manufacturer:  TCI America E10291G

 View more versions of this product

Catalog No. E10291G



Specifications

Specifications

1-Ethoxy-4-ethynylbenzene
1g
C10H10O
4-Ethoxyphenylacetylene
CCOC1=CC=C(C=C1)C#C
146.189
146.19
≥98.0% (GC)
Yellow
79887-14-2
MFCD00173880
FRGNOZUOTHMJSC-UHFFFAOYSA-N
1-ethoxy-4-ethynylbenzene
2775124
Clear Liquid at 20°C

Chemical Identifiers

79887-14-2
146.189
FRGNOZUOTHMJSC-UHFFFAOYSA-N
2775124
CCOC1=CC=C(C=C1)C#C
C10H10O
MFCD00173880
4-Ethoxyphenylacetylene
1-ethoxy-4-ethynylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents